ENA444725220 Enamine

3-(1-methyl-1H-pyrazol-5-yl)-8-azabicyclo[3.2.1]oct-2-ene

  • MDL number MFCD28348293
  • Cas Number : 1692516-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204129
Availability details for
Quantity
  • Availability
  • Add to Cart