ENA444725223 Enamine

4-(3,5-dimethoxyphenyl)-4-fluoropiperidine hydrochloride

  • MDL number MFCD28348296
  • Cas Number : 1803603-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204172
Availability details for
Quantity
  • Availability
  • Add to Cart