ENA444725227 Enamine

3-(3,5-difluorophenyl)-3-fluoropyrrolidine hydrochloride

  • MDL number MFCD28348300
  • Cas Number : 1803583-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204201
Availability details for
Quantity
  • Availability
  • Add to Cart