ENA444725228 Enamine

4-(5-bromopyridin-2-yl)-1,3-thiazol-2-amine dihydrobromide

  • MDL number MFCD28348301
  • Cas Number : 1803595-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204204
Availability details for
Quantity
  • Availability
  • Add to Cart