ENA444725231 Enamine

2-{[1-(2-fluorophenyl)ethyl]amino}-3-methylbutan-1-ol hydrochloride

  • MDL number MFCD28348304
  • Cas Number : 1803583-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204283
Availability details for
Quantity
  • Availability
  • Add to Cart