ENA444725233 Enamine

methyl 1-[hydroxy(1-methyl-1H-imidazol-2-yl)methyl]cyclopropane-1-carboxylate

  • MDL number MFCD28348306
  • Cas Number : 1803586-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204296
Availability details for
Quantity
  • Availability
  • Add to Cart