ENA444725240 Enamine

1-amino-3,3-dimethylcyclopentane-1-carboxylic acid hydrochloride

  • MDL number MFCD28348314
  • Cas Number : 1803601-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204432
Availability details for
Quantity
  • Availability
  • Add to Cart