ENA444725245 Enamine

[1-(butane-1-sulfonyl)pyrrolidin-3-yl]methanamine hydrochloride

  • MDL number MFCD28348319
  • Cas Number : 1803587-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204549
Availability details for
Quantity
  • Availability
  • Add to Cart