ENA444725247 Enamine

N-{2-[(thian-4-yl)amino]ethyl}acetamide hydrochloride

  • MDL number MFCD28348322
  • Cas Number : 1803611-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204574
Availability details for
Quantity
  • Availability
  • Add to Cart