ENA444725276 Enamine

N-{[4-bromo-2-(trifluoromethyl)phenyl]methyl}-3-methylpyridin-2-amine

  • MDL number MFCD28348349
  • Cas Number : 1803582-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61448
Availability details for
Quantity
  • Availability
  • Add to Cart