ENA444725284 Enamine

1-(methylsulfanyl)cyclobutane-1-carbaldehyde

  • MDL number MFCD24679687
  • Cas Number : 35120-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202898
Availability details for
Quantity
  • Availability
  • Add to Cart