ENA444725289 Enamine

2-{4-[(dimethylamino)methyl]phenyl}benzoic acid

  • MDL number MFCD11932172
  • Cas Number : 1159492-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203376
Availability details for
Quantity
  • Availability
  • Add to Cart