ENA444725292 Enamine

({2-[(2-fluorophenyl)methoxy]-3-methoxyphenyl}methyl)(methyl)amine hydrochloride

  • MDL number MFCD07106791
  • Cas Number : 1049713-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203384
Availability details for
Quantity
  • Availability
  • Add to Cart