ENA444725295 Enamine

2-(4-methylcyclohexyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD21323426
  • Cas Number : 1462882-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202920
Availability details for
Quantity
  • Availability
  • Add to Cart