ENA444725297 Enamine

ethyl 2-phenyl-1H-imidazole-4-carboxylate

  • MDL number MFCD03426222
  • Cas Number : 32683-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203391
Availability details for
Quantity
  • Availability
  • Add to Cart