ENA444725299 Enamine

2-chloropropan-1-amine hydrochloride

  • MDL number MFCD18976962
  • Cas Number : 6266-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-183
Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202926
Availability details for
Quantity
  • Availability
  • Add to Cart