ENA444725302 Enamine

2-oxo-1H,2H,3H-imidazo[4,5-b]pyridine-5-carboxylic acid

  • MDL number MFCD28348220
  • Cas Number : 1783316-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203402
Availability details for
Quantity
  • Availability
  • Add to Cart