ENA444725304 Enamine

3-[(4-acetylpiperazin-1-yl)sulfonyl]-4-fluorobenzoic acid

  • MDL number MFCD16559494
  • Cas Number : 1053994-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203406
Availability details for
Quantity
  • Availability
  • Add to Cart