ENA444725307 Enamine

1-(2-cyclopropyl-1,3-oxazol-4-yl)ethan-1-one

  • MDL number MFCD28348223
  • Cas Number : 1519375-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203448
Availability details for
Quantity
  • Availability
  • Add to Cart