ENA444725309 Enamine

2-(3-methyl-2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)acetic acid

  • MDL number MFCD11643032
  • Cas Number : 10490-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203453
Availability details for
Quantity
  • Availability
  • Add to Cart