ENA444725310 Enamine

3-(3-fluorophenyl)prop-2-yn-1-amine hydrochloride

  • MDL number MFCD25458181
  • Cas Number : 1439904-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202929
Availability details for
Quantity
  • Availability
  • Add to Cart