ENA444725316 Enamine

5-(6-methoxypyridin-3-yl)-2-methylaniline

  • MDL number MFCD16103986
  • Cas Number : 1248688-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203468
Availability details for
Quantity
  • Availability
  • Add to Cart