ENA444725317 Enamine

5-(1-methyl-1H-pyrazol-4-yl)thiophene-2-carbaldehyde

  • MDL number MFCD22410646
  • Cas Number : 1231192-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203469
Availability details for
Quantity
  • Availability
  • Add to Cart