ENA444725323 Enamine

N-cyclopentyl-N-methylcarbamoyl chloride

  • MDL number MFCD23790130
  • Cas Number : 1383546-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202937
Availability details for
Quantity
  • Availability
  • Add to Cart