ENA444725329 Enamine

tert-butyl 4-[(prop-2-yn-1-yl)carbamoyl]piperidine-1-carboxylate

  • MDL number MFCD10605357
  • Cas Number : 1030181-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203501
Availability details for
Quantity
  • Availability
  • Add to Cart