ENA444725330 Enamine

1-(2,2-dichlorocyclopropyl)ethan-1-one

  • MDL number MFCD18975826
  • Cas Number : 61971-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202940
Availability details for
Quantity
  • Availability
  • Add to Cart