ENA444725331 Enamine

N-{[4-(1H-1,2,4-triazol-1-yl)phenyl]methyl}cyclopropanamine

  • MDL number MFCD11629610
  • Cas Number : 1095118-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203520
Availability details for
Quantity
  • Availability
  • Add to Cart