ENA444725332 Enamine

2-isocyanato-1,1-dimethylcyclopropane

  • MDL number MFCD18269785
  • Cas Number : 1343804-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203522
Availability details for
Quantity
  • Availability
  • Add to Cart