ENA444725338 Enamine

2-bromo-1-[2-(methylsulfanyl)phenyl]ethan-1-one

  • MDL number MFCD23845003
  • Cas Number : 22165-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-82
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203528
Availability details for
Quantity
  • Availability
  • Add to Cart