ENA444725339 Enamine

2-bromo-6-chlorobenzene-1-thiol

  • MDL number MFCD18089361
  • Cas Number : 1263376-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203530
Availability details for
Quantity
  • Availability
  • Add to Cart