ENA444725356 Enamine

5-(aminomethyl)-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD27986908
  • Cas Number : 1638763-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203616
Availability details for
Quantity
  • Availability
  • Add to Cart