ENA444725359 Enamine

5-(4-chloro-3-methylphenoxy)pentan-2-one

  • MDL number MFCD16165536
  • Cas Number : 1225537-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203620
Availability details for
Quantity
  • Availability
  • Add to Cart