ENA444725361 Enamine

2-amino-3-hydroxybenzene-1-sulfonamide

  • MDL number MFCD16999363
  • Cas Number : 847229-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203622
Availability details for
Quantity
  • Availability
  • Add to Cart