ENA444725362 Enamine

3-iodo-1-(3,3,3-trifluoropropyl)-1H-pyrazole

  • MDL number MFCD23768992
  • Cas Number : 1506585-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202976
Availability details for
Quantity
  • Availability
  • Add to Cart