ENA444725371 Enamine

ethyl 2-amino-1,8-naphthyridine-3-carboxylate

  • MDL number MFCD24623616
  • Cas Number : 51527-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203004
Availability details for
Quantity
  • Availability
  • Add to Cart