ENA444725372 Enamine

tert-butyl 2-carbamothioyl-2,2-dimethylacetate

  • MDL number MFCD24625932
  • Cas Number : 75235-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203006
Availability details for
Quantity
  • Availability
  • Add to Cart