ENA444725379 Enamine

2-[(2-bromoethyl)sulfanyl]propane

  • MDL number MFCD12764180
  • Cas Number : 5755-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203661
Availability details for
Quantity
  • Availability
  • Add to Cart