ENA444725383 Enamine

4-bromo-N,N-diethyl-3-fluoroaniline

  • MDL number MFCD20920109
  • Cas Number : 761001-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203669
Availability details for
Quantity
  • Availability
  • Add to Cart