ENA444725384 Enamine

4-cyclopropyl-2-methylaniline

  • MDL number MFCD14585544
  • Cas Number : 1202161-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203700
Availability details for
Quantity
  • Availability
  • Add to Cart