ENA444725385 Enamine

2-(7-methoxy-1-benzofuran-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD18383011
  • Cas Number : 1072811-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203701
Availability details for
Quantity
  • Availability
  • Add to Cart