ENA444725386 Enamine

4-methoxy-N,N-dimethylbutanamide

  • MDL number MFCD18827185
  • Cas Number : 289625-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203702
Availability details for
Quantity
  • Availability
  • Add to Cart