ENA444725390 Enamine

4-(4-bromophenyl)-4-methyl-2-sulfanyl-4,5-dihydro-1H-imidazol-5-one

  • MDL number MFCD24345047
  • Cas Number : 1552483-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203709
Availability details for
Quantity
  • Availability
  • Add to Cart