ENA444725393 Enamine

5-(2,2-difluoroethoxy)-N'-hydroxypyrazine-2-carboximidamide

  • Cas Number : 1564175-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203714
Availability details for
Quantity
  • Availability
  • Add to Cart