ENA444725396 Enamine

5-(3-bromophenyl)-1H-1,2,4-triazole-3-carbonitrile

  • MDL number MFCD01789804
  • Cas Number : 328281-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203724
Availability details for
Quantity
  • Availability
  • Add to Cart