ENA444725413 Enamine

3-amino-N-[2-(dimethylamino)ethyl]-1H-pyrazole-5-carboxamide

  • MDL number MFCD20316253
  • Cas Number : 1347088-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203774
Availability details for
Quantity
  • Availability
  • Add to Cart