ENA444725414 Enamine

2-bromo-6-chloro-4-methylphenol

  • MDL number MFCD22414716
  • Cas Number : 57018-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203031
Availability details for
Quantity
  • Availability
  • Add to Cart