ENA444725419 Enamine

2-[(2-bromoethyl)sulfanyl]-2-methylpropane

  • MDL number MFCD12172032
  • Cas Number : 5755-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203782
Availability details for
Quantity
  • Availability
  • Add to Cart