ENA444725420 Enamine

(2-bromo-5-methoxyphenyl)methanesulfonamide

  • MDL number MFCD21193490
  • Cas Number : 1485213-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203041
Availability details for
Quantity
  • Availability
  • Add to Cart