ENA444725428 Enamine

[3-methanesulfonyl-5-(trifluoromethyl)phenyl]methanol

  • MDL number MFCD26051392
  • Cas Number : 1003843-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202871
Availability details for
Quantity
  • Availability
  • Add to Cart