ENA444725429 Enamine

2-hydroxyethyl N-benzylcarbamate

  • MDL number MFCD00865284
  • Cas Number : 4663-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203841
Availability details for
Quantity
  • Availability
  • Add to Cart